missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ProSci Death Receptor 6 Recombinant Protein
Description
This recombinant protein can be used for biological assays. For research use only.
Tumor Necrosis Factor Receptor Superfamily Member 21 (TNFRSF21) is a type I transmembrane receptor that includes four extracellular cysteine-rich motifs and a cytoplasmic death domain. DR6 is highly expressed in heart, brain, placenta, pancreas, lymph node, thymus and prostate. DR6 may activate NF-kappa-B and JNK to promote apoptosis and T-cell differentiation. In addition, DR6 binds with N-APP, which is released by the deprivation of Trophic-factor. It triggers caspase activation and degeneration of both neuronal cell bodies (via caspase-3) and axons (via caspase-6). DR6 is also expressed on the tumor cell lines and can be induced by TNF- alpha.
Specifications
Specifications
| Accession Number | O75509 |
| For Use With (Application) | Not Available |
| Gene ID (Entrez) | 27242 |
| Molecular Weight (g/mol) | 34.62 kDa |
| Quantity | 0.05 mg |
| Source | Human Cells |
| Common Name | tumor necrosis factor receptor superfamily member 21 |
| Gene Symbol | TNFRSF21 |
| Product Type | Recombinant Protein |
| Conjugate | Unconjugated |
| Show More |