missing translation for 'onlineSavingsMsg'
Learn More

Cytiva DEAE Sepharose™ Fast Flow Chromatography Media

Catalog No. 45002930
Change view
Click to view available options
Packaging Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
45002930 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002930 Supplier Cytiva Supplier No. 17070910

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For preparative protein separations in both research and industrial applications. Cytiva Life Sciences™ DEAE Sepharose™ Fast Flow Chromatography Media can be used for capture and intermediate purification of wide range of approved biopharmaceuticals.

  • BioProcess media
  • Weak anion exchanger
  • High chemical stability allows for well proven CIP and sanitization protocols
  • Composed of crosslinked 6% agarose beads with diethylaminoethyl (DEAE)
  • Hydrophilic nature of base matrix ensures low levels of non-specific binding
  • Average particle size of 90μm
  • pH Stability working range from 2 to 12
  • Pack size of 25mL

Specifications

Quantity 25 mL
For Use With (Application) For Preparative Protein Separations in Both Research and Industrial AppliCatIons
Stability Stable in commonly used aqueous Buffers: 1 M NaOH, 8 M Urea, 8 M Guanidine Hydrochloride, 70% Ethanol
Storage Requirements 4°C to 30°C
Product Type DEAE Sepharose Fast Flow Media
Flow Rate 300 to 600 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 5.9 in.
Height (Metric) Bed 15 cm
Ionic Capacity 0.11 to 0.16 mM Cl-/mL
Ion Exchange Type Weak AnIon Exchanger
Max. Pressure 100 kPa
Matrix 6% Cross-Linked Agarose with Dextran surface extenders
pH Range 2 to 12
Storage Buffer 20% Ethanol
Ligand Type Diethylaminoethyl
Packaging Quantity 25 mL
Show More Show Less