Learn More
MilliporeSigma™ Supelco™ Deactivated (Silanized) Glass Inlet Liner, Unpacked
For Packed Injection Ports
$253.41 - $762.32
Specifications
Deactivation Silanized
Packing Material None
Product Type Inlet Liner
For Use With (Application) Gas Chromatography (GC)
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Quantity Price Quantity & Availability  
111010483
View Documents Promotion Details
MilliporeSigma™ Supelco™
20508
1.8 mm 3.0 mm 5/Pk.
Each for $271.06
Only null left
111008661
View Documents Promotion Details
MilliporeSigma™ Supelco™
20511
1.8 mm 3.0 mm 25/Pk.
Each for $762.32
Only null left
111010515
View Documents Promotion Details
MilliporeSigma™ Supelco™
2631605
3.0 mm 6.0 mm 5/Pk.
Each for $253.41
Only null left
Description
Description
Specifications
Specifications
Silanized
None
Inlet Liner
Gas Chromatography (GC)