missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NoTrax™ De-Flex™
De-Flex™ heavy duty rubber mat is the latest technology in modular flooring systems
Supplier: NoTrax™ 570S1818BL
Description
- Ultimate heavy duty floor mat for durability, comfort and safety
- Rubber formulation with bulleted domes underneath, gives it the ability to bend and flex with every movement
- 18 x 18 in. tiles can create any size floor mat
- Diamond plate traction provides additional safety
- Ideal for industrial floor use
- Commonly used for long runs, custom configurations, or wherever heavy equipment floor mats are needed
Chemical Identifiers
| Black | |
| Heavy duty 0.75 in. rubber | |
| 18 in. |
| 18 in. | |
| Industrial Mats |
Specifications
| Heavy duty 0.75 in. rubber | |
| Industrial Mats | |
| 18 in. |
| Black | |
| 18 in. | |
| 1 |