missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX4 Recombinant Rabbit Monoclonal Antibody (PSH08-53)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA562888
Description
Positive Control: Mouse testis tissue lysate, human testis tissue, mouse testis tissue, mouse ovary tissue.Tissue Specificity: Tissue enriched (testis)Subcellular Location: Cytoplasm, perinuclear region.Sequence Similarities: 92% Mouse/RatPredicted band size: 79 kDa.
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is a homolog of VASA proteins in Drosophila and several other species. The gene is specifically expressed in the germ cell lineage in both sexes and functions in germ cell development. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| DDX4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q61496, Q9NQI0 | |
| DDX4 | |
| Recombinant protein within human DDX4 aa 201-724. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| PSH08-53 | |
| Unconjugated | |
| DDX4 | |
| ATP-dependent RNA helicase DDX4; AV206478; Cvh; Ddx4; D-E-A-D (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (aspartate-glutamate-alanine-aspartate) box polypeptide 4; DEAD (Asp-Glu-Ala-Asp) box polypeptide 4; DEAD box polypeptide 4; DEAD box protein 4; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4; DEAD-box helicase 4; MGC111074; Mvh; mvh / m'vasa; probable ATP-dependent RNA helicase DDX4; putative ATP-dependent RNA helicase DDX4; RVLG; VASA; Vasa homolog; vasa-like gene protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 13206, 54514 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |