missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDX3/DDX3Y Recombinant Rabbit Monoclonal Antibody (1N2I8)
GREENER_CHOICE

Catalog No. MA569170
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA569170 100 μL
MA569171 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA569170 Supplier Invitrogen™ Supplier No. MA569170

Rabbit Recombinant Monoclonal Antibody

DDX3/DDX3Y Recombinant Monoclonal Antibody for ELISA, ICC/IF, IHC (P), IP, Western Blot

DDX3 (DEAD-box helicase 3) is an RNA helicase involved in multiple cellular processes, including RNA splicing, transport, translation, and degradation. It has key roles in antiviral responses, cell cycle regulation, and apoptosis. DDX3 is involved in the regulation of gene expression at multiple levels and is critical in processes such as cell cycle regulation, apoptosis, and innate immune response to viral infections. DDX3 has been shown to interact with various cellular proteins and RNA molecules, influencing their stability and function. DDX3Y, also known as DBY, belongs to the DEAD-box protein family and is located on the Y chromosome at Yq11.It shares significant sequence and functional similarity with DDX3 but has a more restricted expression pattern, predominantly in the testes. Mutations or deletions in DDX3Y can lead to male infertility due to impaired spermatogenesis. The protein encoded by DDX3Y consists of conserved motifs for ATP binding and hydrolysis necessary for RNA unwinding, making it vital for the proper translation and stability of mRNAs during spermatogenesis.

Specifications

Antigen DDX3/DDX3Y
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 1N2I8
Concentration 1.11 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3.
Gene DDX3Y
Gene Accession No. O00571, O15523, Q62095, Q62167
Gene Symbols DDX3X, DDX3Y
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13205, 1654, 26900, 317335, 364073, 8653
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less