missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDX17 Recombinant Rabbit Monoclonal Antibody (23GB3560)
GREENER_CHOICE

Catalog No. MA552566
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552566 100 μL
MA552567 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552566 Supplier Invitrogen™ Supplier No. MA552566
Only null left

Rabbit Recombinant Monoclonal Antibody

DDX17 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and splicesosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which is an ATPase activated by a variety of RNA species, but not by dsDNA. This protein, and that encoded by DDX5 gene, are more closely related to each other than to any other member of the DEAD box family. This gene can encode multiple isoforms due to both alternative splicing and the use of alternative translation initiation codons, including a non-AUG (CUG) start codon.

Specifications

Antigen DDX17
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3560
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DDX17
Gene Accession No. Q501J6, Q92841
Gene Alias 2610007K22Rik; A430025E01Rik; AI047725; C80929; DDX17; DEAD (Asp-Glu-Ala-Asp) box helicase 17; DEAD (Asp-Glu-Ala-Asp) box polypeptide 17; DEAD (Asp-Glu-Ala-Asp) box polypeptide 46; DEAD box polypeptide 17; DEAD box protein 17; DEAD box protein p72; DEAD box protein p82; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 17 (72kD); DEAD/H box 17; DEAD-box helicase 17; DKFZp761H2016; Gm926; P72; Probable ATP-dependent RNA helicase DDX17; RH70; RNA-dependent helicase p72; RP3-434P1.1
Gene Symbols DDX17
Host Species Rabbit
Immunogen A synthetic peptide of human DDX17 (100-174AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10521, 315133, 67040
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less