missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDOST Monoclonal Antibody (OTI2H1)
GREENER_CHOICE

Catalog No. PIMA526609
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526609 Supplier Invitrogen™ Supplier No. MA526609
Only null left

Mouse Monoclonal Antibody

DDOST Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a component of the oligosaccharyltransferase complex which catalyzes the transfer of high-mannose oligosaccharides to asparagine residues on nascent polypeptides in the lumen of the rough endoplasmic reticulum. The protein complex co-purifies with ribosomes. The product of this gene is also implicated in the processing of advanced glycation end products (AGEs), which form from non-enzymatic reactions between sugars and proteins or lipids and are associated with aging and hyperglycemia.

Specifications

Antigen DDOST
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI2H1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DDOST
Gene Accession No. P39656
Gene Alias advanced glycation endproduct receptor 1; advanced glycation end-product receptor 1; AGER1; AGE-R1; CDG1R; Ddost; DDOST 48 kDa subunit; dolichyl-diphosphooligosaccharide protein glycosyltransferase; dolichyl-diphosphooligosaccharide-protein glycosyltransferase; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic); dolichyl-di-phosphooligosaccharide-protein glycotransferase; dolichyl-diphosphooligosaccharide-protein glycotransferase; KIAA0115; MGC2191; OK/SW-cl.45; OKSWcl45; oligosaccharyl transferase 48 kDa subunit; oligosaccharyltransferase 48 kDa subunit; oligosaccharyltransferase OST48; oligosaccharyltransferase subunit 48; OST; OST48; WBP1; wu:fa11d01; zgc:66068; zgc:66068 protein; zgc:77478
Gene Symbols DDOST
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 131-378 of DDOST produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1650
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less