missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDAH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552278
Description
Immunogen sequence: SKRTNQRGAE ILADTFKDYA VSTVPVADGL HLKSFCSMAG PNLIAIGSSE SAQKALKIMQ QMSDHRYDKL TVPDDIAANC IYLNIPNKGH VLLHRTPEEY PESAKVYEKL KDHMLIPVSM SELEKVDGLL T Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity.Specifications
| DDAH1 | |
| Polyclonal | |
| Unconjugated | |
| DDAH1 | |
| 2410006N07Rik; 2510015N06Rik; AI987801; AW050362; Ddah; Ddah1; DDAH-1; DDAHI; dimethylargininase-1; dimethylarginine dimethylaminohydrolase 1; epididymis secretory protein Li 16; FLJ21264; FLJ25539; HEL-S-16; N(G),N(G)-dimethylarginine dimethylaminohydrolase 1; NG, NG-dimethylarginine dimethylaminohydrolase; NG,NG dimethylarginine dimethylaminohydrolase; RP4-621F18.1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23576, 64157, 69219 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O08557, O94760, Q9CWS0 | |
| DDAH1 | |
| Recombinant protein corresponding to Human DDAH1. Recombinant protein control fragment (Product #RP-88775). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |