missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDAH1 Monoclonal Antibody (K24015_11E11), Invitrogen™
Mouse Monoclonal Antibody
$1,461.54 - $2,194.06
Specifications
| Antigen | DDAH1 |
|---|---|
| Clone | K24015_11E11 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: ARAERQHQLY VGVLGSKLGL QVVELPADES LPDCVFVEDV AVVCEETALI TRPGAPSRRK EVDMMKEALE KLQLNIVEMK DENATLDGGD
DDAH1 Monoclonal antibody specifically detects DDAH1 in Human samples. It is validated for Western BlotSpecifications
| DDAH1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 2410006N07Rik; 2510015N06Rik; AI987801; AW050362; Ddah; Ddah1; DDAH-1; DDAHI; dimethylargininase-1; dimethylarginine dimethylaminohydrolase 1; epididymis secretory protein Li 16; FLJ21264; FLJ25539; HEL-S-16; N(G),N(G)-dimethylarginine dimethylaminohydrolase 1; NG, NG-dimethylarginine dimethylaminohydrolase; NG,NG dimethylarginine dimethylaminohydrolase; RP4-621F18.1 | |
| Recombinant human DDAH1 (immunogen range is 41-130). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| DDAH1 |
| K24015_11E11 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O94760 | |
| 23576 | |
| IgG1 | |
| Protein A | |
| Antibody |