missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dcp2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories Inc A302597AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
The removal, or decapping, of eukaryotic mRNA is an important step in the degradation of mRNA. Decapping protein 2 (DCP2) is the major mRNA decapping enzyme in cells. It is a member of the Nudix hydrolases superfamily of proteins that predomitly catalyze the hydrolysis of small nucleoside diphosphate substrates linked to another moiety. DCP2 is widely expressed in multiple tissues at varying levels, with highest expression seen in testis and brain.Specifications
| Dcp2 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q8IU60 | |
| DCP2 | |
| Between 370 and 420 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| DCP2 | |
| NUDT20, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 167227 | |
| 4° C | |
| Liquid |