missing translation for 'onlineSavingsMsg'
Learn More

DCJTB 98.0+%, TCI America™

Catalog No. D4700200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4700200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4700200MG Supplier TCI America Supplier No. D4700200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 200052-70-6
Molecular Formula C30H35N3O
Molecular Weight (g/mol) 453.63
InChI Key HXWWMGJBPGRWRS-CMDGGOBGSA-N
Synonym 2-tert-Butyl-4-(dicyanomethylene)-6-[2-(1,1,7,7-tetramethyljulolidin-9-yl)vinyl]-4H-pyran
PubChem CID 21889100
SMILES CC1(CCN2CCC(C3=C2C1=CC(=C3)C=CC4=CC(=C(C#N)C#N)C=C(O4)C(C)(C)C)(C)C)C

Specifications

Quantity 200 mg
Formula Weight 453.63
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material DCJTB