accessibility menu, dialog, popup

UserName

CAS RN 200052-70-6

IUPAC Name: 2-[2-tert-butyl-6-(2-{4,4,10,10-tetramethyl-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-trien-7-yl}ethenyl)-4H-pyran-4-ylidene]propanedinitrile
Synonyms: 2-[2-tert-butyl-6-(2-{4,4,10,10-tetramethyl-1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-trien-7-yl}ethenyl)pyran-4-ylidene]propanedinitrile
Molecular Weight (g/mol): 453.63
Molecular Formula: C30H35N3O
InChi Key: HXWWMGJBPGRWRS-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(C=C(O1)C=CC1=CC2=C3N(CCC(C)(C)C3=C1)CCC2(C)C)=C(C#N)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results