missing translation for 'onlineSavingsMsg'
Learn More

DBX1 Antibody, ProSci

Catalog No. 16594404
Change view
Click to view available options
Quantity:
20 μg Trial Size
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16594404 20 μg Trial Size
16917709 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16594404 Supplier ProSci Llc Supplier No. 64550020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Specifications

Antigen DBX1
Applications ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution DBX1 antibody can be used for detection of DBX1 by Western blot at 1 - 2 μg/mL. Antibody can also be used for immunohistochemistry starting at 2.5 μg/mL. For immunofluorescence start at 20 μg/mL. Antibody validated: Western Blot in mouse samples; Immunohistochemistry in human samples and Immunofluorescence in human samples. All other applications and species not yet tested.
Formulation DBX1 Antibody is supplied in PBS containing 0.02% sodium azide.
Gene Accession No. NP_001025036
Gene Alias DBX1 Antibody: Homeobox protein DBX1, Developing brain homeobox protein 1
Host Species Rabbit
Purification Method DBX1 Antibody is affinity chromatography purified via peptide column.
Quantity 20 μg Trial Size
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120237
Test Specificity BX1 antibody is predicted to not cross-react with other DBX protein family members
Target Species Human, Mouse, Rat
Content And Storage DBX1 antibody can be stored at 4°C for three months and -20°C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Product Type Antibody
Form Liquid
Isotype IgG
Research Category Stem Cell
Show More Show Less