missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DAZAP1 Recombinant Rabbit Monoclonal Antibody (23GB3540)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA552614
Description
DAZAP1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
The androgen receptor (AR) is a member of the steroid-hormone receptor superfamily of nuclear receptors. The receptor is more than 90 kDa and has three major functional domains: the N-terminal domain, DNA-binding domain, and the androgen-binding domain. The androgen receptor is a ligand-activated transcription factor that binds active testosterone (T) and dihydrotestosterone (DHT). Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. The AR signaling pathway plays a key role in development and function of male reproductive organs, including the prostate and epididymis.AR also plays a role in nonreproductive organs, such as muscle, hair follicles, and brain.Androgen Receptor is a phosphoprotein, and also regulates mitogen-activated protein kinase (MAP kinase). The inhibition of the MEK1/2 pathway correlates directly with a change in phosphorylation state of the androgen receptor. Abnormalities in the AR signaling pathway have been linked to a number of diseases, including prostate cancer, Kennedy's disease, and male infertility. Mutations in this gene are associated with complete androgen insensitivity (CAIS).Chemical Identifiers
| DAZAP1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| DAZAP1 | |
| 2410042M16Rik; DAZ associated protein 1; DAZAP1; DAZ-associated protein 1; Deleted in azoospermia-associated protein 1; LOC561459 protein; LOW QUALITY PROTEIN: DAZ-associated protein 1; MGC19907; mPrrp; testicular tissue protein Li 50 | |
| DAZAP1 | |
| Primary | |
| Affinity chromatography |
| 23GB3540 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96EP5, Q9JII5 | |
| 26528, 362836, 70248 | |
| A synthetic peptide of human DAZAP1 (265-407AA). | |
| Antibody |
Specifications
| DAZAP1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DAZAP1 | |
| 2410042M16Rik; DAZ associated protein 1; DAZAP1; DAZ-associated protein 1; Deleted in azoospermia-associated protein 1; LOC561459 protein; LOW QUALITY PROTEIN: DAZ-associated protein 1; MGC19907; mPrrp; testicular tissue protein Li 50 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26528, 362836, 70248 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3540 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96EP5, Q9JII5 | |
| DAZAP1 | |
| A synthetic peptide of human DAZAP1 (265-407AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |