missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daunorubicin Hydrochloride 98.0+%, TCI America™
Supplier: TCI America D4532100MG
Specifications
| Daunorubicin Hydrochloride | |
| 167°C | |
| MFCD04974507 | |
| 2811 | |
| GUGHGUXZJWAIAS-QQYBVWGSSA-N | |
| (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride | |
| 62770 | |
| ≥98.0% (HPLC,N) |
| 23541-50-6 | |
| C27H30ClNO10 | |
| 100 mg | |
| daunorubicin hydrochloride, daunorubicin hcl, daunomycin hydrochloride, daunoblastina, rubidomycin hydrochloride, daunorubicinol hydrochloride, daunoblastin, rubomycin, ondena, daunomycin chlorohydrate | |
| CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl | |
| 563.984 | |
| 563.98 | |
| Crystalline Powder |
Chemical Identifiers
| 23541-50-6 | |
| 563.984 | |
| GUGHGUXZJWAIAS-QQYBVWGSSA-N | |
| 62770 | |
| CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl |
| C27H30ClNO10 | |
| MFCD04974507 | |
| daunorubicin hydrochloride, daunorubicin hcl, daunomycin hydrochloride, daunoblastina, rubidomycin hydrochloride, daunorubicinol hydrochloride, daunoblastin, rubomycin, ondena, daunomycin chlorohydrate | |
| (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride |
Safety and Handling
RTECSNumber : HB7878000
TSCA : No
Recommended Storage : Refrigerator