missing translation for 'onlineSavingsMsg'
Learn More

Daunorubicin Hydrochloride 98.0+%, TCI America™

Catalog No. p-7128324
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D453220MG 20 mg
D4532100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D453220MG Supplier TCI America Supplier No. D453220MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23541-50-6
Molecular Formula C27H30ClNO10
Molecular Weight (g/mol) 563.984
MDL Number MFCD04974507
InChI Key GUGHGUXZJWAIAS-QQYBVWGSSA-N
Synonym daunorubicin hydrochloride, daunorubicin hcl, daunomycin hydrochloride, daunoblastina, rubidomycin hydrochloride, daunorubicinol hydrochloride, daunoblastin, rubomycin, ondena, daunomycin chlorohydrate
PubChem CID 62770
IUPAC Name (7S,9S)-9-acetyl-7-[(2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyloxan-2-yl]oxy-6,9,11-trihydroxy-4-methoxy-8,10-dihydro-7H-tetracene-5,12-dione;hydrochloride
SMILES CC1C(C(CC(O1)OC2CC(CC3=C(C4=C(C(=C23)O)C(=O)C5=C(C4=O)C=CC=C5OC)O)(C(=O)C)O)N)O.Cl

Specifications

Melting Point 167°C
Quantity 20 mg
UN Number 2811
Formula Weight 563.98
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Daunorubicin Hydrochloride
RTECSNumber HB7878000
TSCA No
Recommended Storage Refrigerator