Learn More
Interscience DataLink
Catalog No. 16949014
Change view
Click to view available options
Product Line:
DataLink
DataLink Pro
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Line
16949014 DataLink
16948922 DataLink Pro
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16949014 Supplier Interscience Supplier No. 410100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The DataLink system transmits, prints and labels the edge of Petri dishes for easy identification.
DataLink makes it easy to identify Petri dishes during its microbiological analysis. The DataLink software controls and transmits the data on a label. All the information relating to the analysis of the sample is traced and automated.
  • User account management
  • Monitoring of sample data and automation of settings
  • Audit trail, 21 CFR part 11
DataLink:
  • Label size: 50 x 10 mm
DataLink Pro:
  • Label size: 50 x 5 mm
  • Semi-automatic labelling platform
Specifications
For Use With (Application) Labeling Petri Dishes
Software DataLink
Product Line DataLink
Voltage 100/220 V