missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ DAR-4M, Calbiochem™,

Catalog No. 2517601MG
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2517601MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2517601MG Supplier MilliporeSigma™ Supplier No. 2517601MG

A rhodamine-based photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of approx. 10nM

Chemical Identifiers

CAS 339527-79-6
Molecular Formula C25H26N4O3
Molecular Weight (g/mol) 430.51
InChI Key SEHXCXYNFKCWQY-UHFFFAOYSA-N
Synonym diaminorhodamine-4m, dar-4m, 3,6-bis dimethylamino-9-3-amino-4-n-methylamino-2-carboxyphenyl xanthylium, dar-4m solution 5 mm in dmso , 1 mg in 0.46 ml dmso
PubChem CID 9980126

Specifications

Color Red-Violet
Quantity 1 mg
Formula Weight 430.5
Grade HPLC
Physical Form Liquid
Chemical Name or Material DAR-4M
Recommended Storage -20°C (-4°F)