accessibility menu, dialog, popup

UserName

CH3NNCH3CH3ONCH3H3CH2NOHO(E/Z)

CAS RN 339527-79-6

IUPAC Name: 2-amino-6-[3,6-bis(dimethylamino)-9H-xanthen-9-ylidene]-3-(methylimino)cyclohexa-1,4-diene-1-carboxylic acid
Synonyms: 2-amino-6-[3,6-bis(dimethylamino)xanthen-9-ylidene]-3-(methylimino)cyclohexa-1,4-diene-1-carboxylic acid
Molecular Weight (g/mol): 430.51
Molecular Formula: C25H26N4O3
InChi Key: SEHXCXYNFKCWQY-UHFFFAOYSA-N
SMILES: CN=C1C=CC(C(C(O)=O)=C1N)=C1C2=C(OC3=C1C=CC(=C3)N(C)C)C=C(C=C2)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results