missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Daptomycin

Catalog No. AC461371000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461371000 100 mg
AC461375000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461371000 Supplier Thermo Scientific Chemicals Supplier No. 461371000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Daptomycin, CAS # 103060-53-3, is a semi-synthetic polypeptide antibiotic that has bactericidal activity against vancomycin-resistant gram-positive bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Daptomycin is an antibiotic that activity against vancomycin-resistant gram-positive bacteria including enterococci and Staphylococcus aureus

Applications

• It has bactericidal activity against vancomycin-resistant gram-positive bacteria

• It can bind the bacterial cell membrane causing disruption of the membrane and increasing efflux of potassium ions from bacterial cells, disrupting protein, DNA, and RNA synthesis

Chemical Identifiers

CAS 103060-53-3
Molecular Formula C72H101N17O26
Molecular Weight (g/mol) 1620.67
InChI Key DOAKLVKFURWEDJ-AVSAZHKKSA-N
Synonym daptomycin
PubChem CID 133640189
SMILES CCCCCCCCCC(=O)NC(CC1=CNC2=CC=CC=C21)C(=O)NC(CC(=O)N)C(=O)NC(CC(=O)O)C(=O)NC3C(OC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC(=O)C(NC(=O)C(NC(=O)C(NC(=O)C(NC(=O)CNC3=O)CCCN)CC(=O)O)C)CC(=O)O)CO)C(C)CC(=O)O)CC(=O)C4=CC=CC=C4N)C

Specifications

Loss on Drying 10% max.
Infrared Spectrum Authentic
Quantity 100 mg
Solubility Information (0.5% in MeOH) Clear colorless to light yellow
Formula Weight 1620.67
Percent Purity ≥93% (HPLC)
Chemical Name or Material Daptomycin

RUO – Research Use Only