missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dapoxetine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America D47615G
Specifications
| Dapoxetine Hydrochloride | |
| 181°C | |
| C21H24ClNO | |
| 5 g | |
| IHWDIQRWYNMKFM-BDQAORGHSA-N | |
| (1S)-N,N-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine;hydrochloride | |
| 71352 | |
| ≥98.0% (HPLC) |
| 129938-20-1 | |
| White-Yellow | |
| MFCD08272809 | |
| dapoxetine hydrochloride, dapoxetine hcl, priligy, ly 210448 hydrochloride, unii-u4oht63mri, u4oht63mri, ly-210448 hydrochloride, dapoxetine hydrochloride usan, s-n,n-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine hydrochloride | |
| CN(C)C(CCOC1=CC=CC2=CC=CC=C21)C3=CC=CC=C3.Cl | |
| 341.879 | |
| 341.88 | |
| Crystalline Powder |
Chemical Identifiers
| 129938-20-1 | |
| 341.879 | |
| IHWDIQRWYNMKFM-BDQAORGHSA-N | |
| 71352 | |
| CN(C)C(CCOC1=CC=CC2=CC=CC=C21)C3=CC=CC=C3.Cl |
| C21H24ClNO | |
| MFCD08272809 | |
| dapoxetine hydrochloride, dapoxetine hcl, priligy, ly 210448 hydrochloride, unii-u4oht63mri, u4oht63mri, ly-210448 hydrochloride, dapoxetine hydrochloride usan, s-n,n-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine hydrochloride | |
| (1S)-N,N-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine;hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator