missing translation for 'onlineSavingsMsg'
Learn More

Dapoxetine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7128446
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47611G 1 g
D47615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47611G Supplier TCI America Supplier No. D47611G
Only null left

Chemical Identifiers

CAS 129938-20-1
Molecular Formula C21H24ClNO
Molecular Weight (g/mol) 341.879
MDL Number MFCD08272809
InChI Key IHWDIQRWYNMKFM-BDQAORGHSA-N
Synonym dapoxetine hydrochloride, dapoxetine hcl, priligy, ly 210448 hydrochloride, unii-u4oht63mri, u4oht63mri, ly-210448 hydrochloride, dapoxetine hydrochloride usan, s-n,n-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine hydrochloride
PubChem CID 71352
IUPAC Name (1S)-N,N-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine;hydrochloride
SMILES CN(C)C(CCOC1=CC=CC2=CC=CC=C21)C3=CC=CC=C3.Cl

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 1 g
Formula Weight 341.88
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dapoxetine Hydrochloride
TSCA No
Recommended Storage Refrigerator