missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DAPK3 (ZIPK), GST Tag Recombinant Protein

Catalog No. PV3686
Encompass
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3686 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV3686 Supplier Invitrogen™ Supplier No. PV3686
Only null left

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

DAPK3 is a serine/threonine kinase which is functionally linked to cell motility, smooth muscle contractility and cell death processes. DAPK3 is encoded by ZIPK and is a component of desmosomes and of the epidermal cornified envelope in keratinocytes. The N-terminal domain of this protein interacts with the plasma membrane and its C-terminus interacts with intermediate filaments. Through its rod domain, this protein forms complexes with envoplakin. This protein may serve as a link between the cornified envelope and desmosomes as well as intermediate filaments. AKT1/PKB, a protein kinase mediating a variety of cell growth and survival signaling processes, is reported to interact with this protein, suggesting a possible role for this protein as a localization signal in AKT1-mediated signaling.

Specifications

Accession Number O43293
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 50% glycerol, 0.02% Triton X-100, 2mM DTT, 150mM NaCl, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 1613
Molecular Weight (g/mol) 80 kDa
Name Human DAPK3 (ZIPK), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias DAP kinase 3; DAP3K; Dapk3; Dapkl; DAP-like kinase; death associated protein kinase 3; death-associated kinase 3; Death-associated like kinase; death-associated protein kinase 3; DLK; MYPT1 kinase; ZIP; ZIP kinase; ZIP kinase isoform; Zipk; ZIP-kinase; zipper-interacting protein kinase
Common Name ZIPK
Gene Symbol DAPK3
KinaseFamily DAPK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody)
Kinase Group Ser⁄Thr Kinases: CAMK Group
Shipping Condition Dry Ice
System Compatibility Z'-LYTE™, Omnia™ Bulk Substrate
Target Entry DAPK3
Target Gene death-associated protein kinase 3
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less