missing translation for 'onlineSavingsMsg'
Learn More

Dansylamide 98.0+%, TCI America™

Catalog No. D15075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15075G 5 g
D15071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D15075G Supplier TCI America Supplier No. D15075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1431-39-6
Molecular Formula C12H14N2O2S
Molecular Weight (g/mol) 250.316
MDL Number MFCD00004000
InChI Key TYNBFJJKZPTRKS-UHFFFAOYSA-N
Synonym dansylamide, dansyl amide, 5-dimethylamino naphthalene-1-sulfonamide, 5-dimethylamino-1-naphthalenesulfonamide, 1-naphthalenesulfonamide, 5-dimethylamino, 1okl, 5-dimethylamino naphthalene-1-sulphonamide, d0s7nh
PubChem CID 65077
IUPAC Name 5-(dimethylamino)naphthalene-1-sulfonamide
SMILES CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)N

Specifications

Melting Point 219°C
Color White-Yellow
Quantity 5 g
Formula Weight 250.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dansylamide [for Fluorometry]