missing translation for 'onlineSavingsMsg'
Learn More

Dansyl-L-phenylalanine 98.0+%, TCI America™

Catalog No. p-7126753
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1500100MG 100 mg
D15001G 1 g
D15005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1500100MG Supplier TCI America Supplier No. D1500100MG

Chemical Identifiers

CAS 1104-36-5
Molecular Formula C21H22N2O4S
Molecular Weight (g/mol) 398.477
MDL Number MFCD00037730
InChI Key GPIOGTIFRDHWSB-SFHVURJKSA-N
Synonym 5-Dimethylaminonaphthalene-1-sulfonyl-L-phenylalanine, Dns-Phe-OH
PubChem CID 13734199
IUPAC Name (2S)-2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]-3-phenylpropanoic acid
SMILES CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)NC(CC3=CC=CC=C3)C(=O)O

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 100 mg
Formula Weight 398.48
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dansyl-L-phenylalanine