CAS RN 1104-36-5
IUPAC Name:
2-[5-(dimethylamino)naphthalene-1-sulfonamido]-3-phenylpropanoic acid
Synonyms:
2-[5-(dimethylamino)naphthalene-1-sulfonamido]-3-phenylpropanoic acid
Molecular Weight (g/mol):
398.48
Molecular Formula:
C21H22N2O4S
InChi Key:
GPIOGTIFRDHWSB-UHFFFAOYNA-N
SMILES:
CN(C)C1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NC(CC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
Dansyl-L-phenylalanine 98.0+%, TCI America™
CAS: 1104-36-5 Molecular Formula: C21H22N2O4S Molecular Weight (g/mol): 398.477 MDL Number: MFCD00037730 InChI Key: GPIOGTIFRDHWSB-SFHVURJKSA-N Synonym: 5-Dimethylaminonaphthalene-1-sulfonyl-L-phenylalanine, Dns-Phe-OH PubChem CID: 13734199 IUPAC Name: (2S)-2-[[5-(dimethylamino)naphthalen-1-yl]sulfonylamino]-3-phenylpropanoic acid SMILES: CN(C)C1=CC=CC2=C1C=CC=C2S(=O)(=O)NC(CC3=CC=CC=C3)C(=O)O