missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dansyl-L-leucine 98.0+%, TCI America™
$67.38 - $329.20
Chemical Identifiers
| CAS | 1100-22-7 |
|---|---|
| Molecular Formula | C18H24N2O4S |
| Molecular Weight (g/mol) | 364.46 |
| MDL Number | MFCD00069532 |
| InChI Key | XBMQRPDOXIPUFG-HNNXBMFYSA-N |
| Synonym | 5-Dimethylaminonaphthalene-1-sulfonyl-L-leucine, Dns-Leu-OH |
| PubChem CID | 150881 |
| IUPAC Name | (2S)-2-[5-(dimethylamino)naphthalene-1-sulfonamido]-4-methylpentanoic acid |
| SMILES | CC(C)C[C@H](NS(=O)(=O)C1=C2C=CC=C(N(C)C)C2=CC=C1)C(O)=O |
Chemical Identifiers
| 1100-22-7 | |
| 364.46 | |
| XBMQRPDOXIPUFG-HNNXBMFYSA-N | |
| 150881 | |
| CC(C)C[C@H](NS(=O)(=O)C1=C2C=CC=C(N(C)C)C2=CC=C1)C(O)=O |
| C18H24N2O4S | |
| MFCD00069532 | |
| 5-Dimethylaminonaphthalene-1-sulfonyl-L-leucine, Dns-Leu-OH | |
| (2S)-2-[5-(dimethylamino)naphthalene-1-sulfonamido]-4-methylpentanoic acid |
Specifications
| 1100-22-7 | |
| C18H24N2O4S | |
| 100 mg | |
| XBMQRPDOXIPUFG-HNNXBMFYSA-N | |
| (2S)-2-[5-(dimethylamino)naphthalene-1-sulfonamido]-4-methylpentanoic acid | |
| 150881 | |
| ≥98.0% (T) | |
| Dansyl-L-leucine |
| 126°C | |
| MFCD00069532 | |
| 5-Dimethylaminonaphthalene-1-sulfonyl-L-leucine, Dns-Leu-OH | |
| CC(C)C[C@H](NS(=O)(=O)C1=C2C=CC=C(N(C)C)C2=CC=C1)C(O)=O | |
| 364.46 | |
| 364.46 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes