accessibility menu, dialog, popup

UserName

CH3H3C(S)NHSOONH3CCH3OHO

CAS RN 1100-22-7

IUPAC Name: (2S)-2-[5-(dimethylamino)naphthalene-1-sulfonamido]-4-methylpentanoic acid
Synonyms: (2S)-2-[5-(dimethylamino)naphthalene-1-sulfonamido]-4-methylpentanoic acid
Molecular Weight (g/mol): 364.46
Molecular Formula: C18H24N2O4S
InChi Key: XBMQRPDOXIPUFG-HNNXBMFYSA-N
SMILES: CC(C)C[C@H](NS(=O)(=O)C1=C2C=CC=C(N(C)C)C2=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results