missing translation for 'onlineSavingsMsg'
Learn More

Dansyl-L-leucine 98.0+%, TCI America™

Catalog No. D1498100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1498100MG 100 mg
D14981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1498100MG Supplier TCI America Supplier No. D1498100MG
Only null left

Chemical Identifiers

CAS 1100-22-7
Molecular Formula C18H24N2O4S
Molecular Weight (g/mol) 364.46
MDL Number MFCD00069532
InChI Key XBMQRPDOXIPUFG-HNNXBMFYSA-N
Synonym 5-Dimethylaminonaphthalene-1-sulfonyl-L-leucine, Dns-Leu-OH
PubChem CID 150881
IUPAC Name (2S)-2-[5-(dimethylamino)naphthalene-1-sulfonamido]-4-methylpentanoic acid
SMILES CC(C)C[C@H](NS(=O)(=O)C1=C2C=CC=C(N(C)C)C2=CC=C1)C(O)=O

Specifications

Melting Point 126°C
Quantity 100 mg
Formula Weight 364.46
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dansyl-L-leucine