missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dansyl Hydrazine 97.0+%, TCI America™
$177.08 - $704.62
Chemical Identifiers
| CAS | 33008-06-9 |
|---|---|
| Molecular Formula | C12H15N3O2S |
| Molecular Weight (g/mol) | 265.33 |
| MDL Number | MFCD00003986 |
| InChI Key | KPQYDVAFRDWIBW-UHFFFAOYSA-N |
| Synonym | 5-Dimethylaminonaphthalene-1-sulfonyl Hydrazine |
| PubChem CID | 94442 |
| IUPAC Name | 5-(dimethylamino)naphthalene-1-sulfonohydrazide |
| SMILES | CN(C)C1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NN |
Chemical Identifiers
| 33008-06-9 | |
| 265.33 | |
| KPQYDVAFRDWIBW-UHFFFAOYSA-N | |
| 94442 | |
| CN(C)C1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NN |
| C12H15N3O2S | |
| MFCD00003986 | |
| 5-Dimethylaminonaphthalene-1-sulfonyl Hydrazine | |
| 5-(dimethylamino)naphthalene-1-sulfonohydrazide |
Specifications
| 33008-06-9 | |
| MFCD00003986 | |
| 5-Dimethylaminonaphthalene-1-sulfonyl Hydrazine | |
| CN(C)C1=C2C=CC=C(C2=CC=C1)S(=O)(=O)NN | |
| 265.33 | |
| 265.33 | |
| Crystalline Powder |
| C12H15N3O2S | |
| 1 g | |
| KPQYDVAFRDWIBW-UHFFFAOYSA-N | |
| 5-(dimethylamino)naphthalene-1-sulfonohydrazide | |
| 94442 | |
| ≥97.0% (T) | |
| Dansyl Hydrazine [for HPLC Labeling] |
Safety and Handling
TSCA : No