missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dansyl Chloride 98.0+%, TCI America™
Supplier: TCI America D06561G
Specifications
| Dansyl Chloride | |
| 70°C | |
| MFCD00003985 | |
| 3261 | |
| XPDXVDYUQZHFPV-UHFFFAOYSA-N | |
| 5-(dimethylamino)naphthalene-1-sulfonyl chloride | |
| 11801 | |
| 269.74 | |
| Crystalline Powder |
| 605-65-2 | |
| C12H12ClNO2S | |
| 1 g | |
| dansyl chloride, 5-dimethylamino naphthalene-1-sulfonyl chloride, dns chloride, dansyl, 5-dimethylamino-1-naphthalenesulfonyl chloride, 1-naphthalenesulfonyl chloride, 5-dimethylamino, 1-dimethylaminonaphthalene-5-sulfonyl chloride, 1-chlorosulfonyl-5-dimethylaminonaphthalene, unii-qmu9166tj4, 5-dimethylaminonaphthalene-1-sulphonyl chloride | |
| CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O | |
| 269.74 | |
| CHEBI:51907 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 605-65-2 | |
| 269.74 | |
| XPDXVDYUQZHFPV-UHFFFAOYSA-N | |
| 11801 | |
| 5-(dimethylamino)naphthalene-1-sulfonyl chloride |
| C12H12ClNO2S | |
| MFCD00003985 | |
| dansyl chloride, 5-dimethylamino naphthalene-1-sulfonyl chloride, dns chloride, dansyl, 5-dimethylamino-1-naphthalenesulfonyl chloride, 1-naphthalenesulfonyl chloride, 5-dimethylamino, 1-dimethylaminonaphthalene-5-sulfonyl chloride, 1-chlorosulfonyl-5-dimethylaminonaphthalene, unii-qmu9166tj4, 5-dimethylaminonaphthalene-1-sulphonyl chloride | |
| CHEBI:51907 | |
| CN(C)C1=CC=CC2=C(C=CC=C12)S(Cl)(=O)=O |
Safety and Handling
RTECSNumber : QK3688000
TSCA : Yes
Recommended Storage : Refrigerator