missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Accuform Signs Danger: Flammable Liquids
Supplier: Accuform Signs MCHG102VS
Specifications
| ANSI Z535, OSHA 29 CFR 1910.145 | |
| 1 Ea. | |
| 14 in. | |
| -20°F | |
| 10 in. | |
| -20°F to 146°F | |
| Chemical Hazard Sign |
| Vinyl | |
| Indoor or short-term outdoor self-stick application | |
| 146°F | |
| 4 mil | |
| -29°C to 63°C | |
| DANGER Flammable Liquid | |
| Red on White |