Learn More
Brady™ DANGER DO NOT OPERATE Perforated Lockout Tags - Two-Part Tag with Detachable Key Stub
Brady™ Perforated Lockout Tags - Two-Part Tag with Detachable Key Stub, Write-On Paper Cardstock: Two-part tags enhance operational control and record-keeping by enabling workers to keep the stubs after attaching the tags, then later give them to managers for verification.
Supplier:  Brady™ WKT3
Catalog No. 19113295
Only null left
Description
Description
  • The excellent write-on surface of B-853 Cardstock is ideal for adding on-the-spot customized notes and other information
  • Paper cardstock tags are recommended for indoor use only
  • 1 in. Dia. metal ring provided for attaching stub to key
  • Not recommended for chemical exposure
Specifications
Specifications
Tag
Cardstock
5.75 x 3 in.
5.75 in.
4.4 in.
Black/Red on White
ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145; OSHA 1910.147
DANGER DO NOT OPERATE
25 Pk.
111.76 mm
Product Suggestions
Product Suggestions