missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Daminozide 98.0+%, TCI America™
$109.98 - $397.05
Chemical Identifiers
| CAS | 1596-84-5 |
|---|---|
| Molecular Formula | C6H12N2O3 |
| Molecular Weight (g/mol) | 160.17 |
| MDL Number | MFCD00002787 |
| InChI Key | NOQGZXFMHARMLW-UHFFFAOYSA-N |
| Synonym | daminozide, kylar, alar, dmasa, aminozide, n-dimethylamino succinamic acid, sadh, b-nine, dimas, succinic acid 2,2-dimethylhydrazide |
| PubChem CID | 15331 |
| ChEBI | CHEBI:4312 |
| IUPAC Name | 3-(N',N'-dimethylhydrazinecarbonyl)propanoic acid |
| SMILES | CN(C)NC(=O)CCC(O)=O |
Chemical Identifiers
| 1596-84-5 | |
| 160.17 | |
| NOQGZXFMHARMLW-UHFFFAOYSA-N | |
| 15331 | |
| 3-(N',N'-dimethylhydrazinecarbonyl)propanoic acid |
| C6H12N2O3 | |
| MFCD00002787 | |
| daminozide, kylar, alar, dmasa, aminozide, n-dimethylamino succinamic acid, sadh, b-nine, dimas, succinic acid 2,2-dimethylhydrazide | |
| CHEBI:4312 | |
| CN(C)NC(=O)CCC(O)=O |
Specifications
| 1596-84-5 | |
| White | |
| C6H12N2O3 | |
| 5 g | |
| NOQGZXFMHARMLW-UHFFFAOYSA-N | |
| 3-(N',N'-dimethylhydrazinecarbonyl)propanoic acid | |
| 15331 | |
| 160.17 | |
| Crystalline Powder |
| 155°C | |
| 3.8 (5% H2O) | |
| MFCD00002787 | |
| daminozide, kylar, alar, dmasa, aminozide, n-dimethylamino succinamic acid, sadh, b-nine, dimas, succinic acid 2,2-dimethylhydrazide | |
| CN(C)NC(=O)CCC(O)=O | |
| 160.17 | |
| CHEBI:4312 | |
| ≥98.0% (GC,T) | |
| Daminozide |
Safety and Handling
RTECSNumber : WM9625000
TSCA : No