accessibility menu, dialog, popup

UserName

CAS RN 1596-84-5

H3CNCH3NHOOHO

CAS RN 1596-84-5

IUPAC Name: 3-(N',N'-dimethylhydrazinecarbonyl)propanoic acid
Synonyms: daminozide
Molecular Weight (g/mol): 160.17
Molecular Formula: C6H12N2O3
InChi Key: NOQGZXFMHARMLW-UHFFFAOYSA-N
SMILES: CN(C)NC(=O)CCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results