missing translation for 'onlineSavingsMsg'
Learn More

Daidzin 98.0+%, TCI America™

Catalog No. D392025MG
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D392025MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D392025MG Supplier TCI America Supplier No. D392025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 552-66-9
Molecular Formula C21H20O9
Molecular Weight (g/mol) 416.38
MDL Number MFCD00017466
InChI Key KYQZWONCHDNPDP-QNDFHXLGSA-N
Synonym daidzin, daidzoside, daidzein 7-o-glucoside, daidzein 7-glucoside, daidzein-7-glucoside, daidzein 7-o-beta-d-glucoside, unii-4r2x91a5m5, daidzein-7-o-glucoside, 7-beta-d-glucopyranosyloxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 7-o-b-d-glucopyranoside
PubChem CID 107971
ChEBI CHEBI:42202
IUPAC Name 3-(4-hydroxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
SMILES OC[C@H]1O[C@@H](OC2=CC=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 242°C
Color White-Yellow
Quantity 25 mg
Formula Weight 416.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Daidzin
RTECSNumber DJ3094000
TSCA No
Recommended Storage Refrigerator