missing translation for 'onlineSavingsMsg'
Learn More

Daidzin, 98%

Catalog No. AC460421000
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460421000 100 mg
AC460420010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460421000 Supplier Thermo Scientific Chemicals Supplier No. 460421000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 552-66-9 | C21H20O9 | 416.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 552-66-9
Molecular Formula C21H20O9
Molecular Weight (g/mol) 416.38
MDL Number MFCD00017466
InChI Key KYQZWONCHDNPDP-QNDFHXLGSA-N
Synonym daidzin, daidzoside, daidzein 7-o-glucoside, daidzein 7-glucoside, daidzein-7-glucoside, daidzein 7-o-beta-d-glucoside, unii-4r2x91a5m5, daidzein-7-o-glucoside, 7-beta-d-glucopyranosyloxy-3-4-hydroxyphenyl-4h-1-benzopyran-4-one, 7-o-b-d-glucopyranoside
PubChem CID 107971
ChEBI CHEBI:42202
IUPAC Name 3-(4-hydroxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxychromen-4-one
SMILES OC[C@H]1O[C@@H](OC2=CC=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Loss on Drying 2.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 mg
Formula Weight 416.38
Percent Purity 98%
Chemical Name or Material Daidzin

RUO – Research Use Only