missing translation for 'onlineSavingsMsg'
Learn More

Daidzein, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014293
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014293 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014293 Supplier Merck Emd Millipore Supplier No. 1658710MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Daidzein is an endocrine-active estrogenic isoflavone. This phytoestrogen is capable to bind to estrogen to exert estrogenic effects in vivo. It can naturally be found in soybean seed and flour.

Featured Industries: Food and Beverages

Specifications

CAS 486-66-8
Physical Form Neat
Linear Formula C15H10O4
MDL Number MFCD00016954
Synonym 4′,7-Dihydroxy­iso­flavone; 7-Hydroxy-3-(4-hydroxy­phenyl)-4 H-1-benzo­pyran-4-one; 7-Hydroxy-3-(4-hydroxy­phenyl)­chromone; Isoaurostatin
Type Analytical Standard
Quantity 10 mg
Formula Weight 254.24
Percent Purity ≥97% (HPLC)
Grade Analytical Standard