missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ DAF-2, Calbiochem™,

Catalog No. 2515001mg
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2515001MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2515001MG Supplier MilliporeSigma™ Supplier No. 2515001MG
Only null left

A highly sensitive and specific fluorescent indicator for the direct detection of nitric oxide (NO) in vitro

Chemical Identifiers

CAS 205391-01-1
Molecular Formula C20H14N2O5
Molecular Weight (g/mol) 362.34
MDL Number MFCD05865100
InChI Key LTYUPYUWXRTNFQ-UHFFFAOYSA-N
Synonym daf-2, 4,5-diaminofluorescein, 5,6-diaminofluorescein, 4,5-diaminofluorescein-isopropanol adduct 1:2, 5,6-diamino-3',6'-dihydroxyspiro 2-benzofuran-1,9'-xanthene-3-one, daf-2 solution 5 mm in dmso , 1 mg in 0.55 ml dmso, 5,6-diamino-3',6'-dihydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, 5,6-diamino-3',6'-dihydroxyspiro 2-benzofuran-1,9'-xanthen-3-one
PubChem CID 10666340
IUPAC Name 5,6-diamino-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES NC1=C(N)C=C2C(=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2

Specifications

Color Yellow
Density 1.69g/cm3 (20°C)
Boiling Point 189°C
Flash Point 87°C (189°F)
Quantity 1 mg
Formula Weight 362.3
Percent Purity ≥98%
Grade HPLC
Concentration or Composition (by Analyte or Components) 5mM solution in DMSO
Physical Form Liquid
Chemical Name or Material DAF-2
Show More Show Less
Recommended Storage -20°C (-4°F); Tightly closed. Transport with dry ice.