accessibility menu, dialog, popup

UserName

OHOOOH2NH2NHO

CAS RN 205391-01-1

IUPAC Name: 5,6-diamino-3',6'-dihydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 5,6-diamino-3',6'-dihydroxyspiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 362.34
Molecular Formula: C20H14N2O5
InChi Key: LTYUPYUWXRTNFQ-UHFFFAOYSA-N
SMILES: NC1=C(N)C=C2C(=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results