missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dabcyl, SE, 4-((4-(dimethylamino)phenyl)azo)benzoic Acid, Succinimidyl Ester

Catalog No. D2245
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2245 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2245 Supplier Invitrogen™ Supplier No. D2245

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

4-((4-(dimethylamino)phenyl)azo)benzoic acid, succinimidyl ester (dabcyl, SE)

The amine-reactive quencher, dabcyl succinimidyl ester has a broad and intense visible absorption but no fluorescence making it useful as an acceptor in fluorescence resonance energy transfer (FRET) applications.

Specifications

Quantity 100 mg
Product Type Dabcyl Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Dabcyl
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.