missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ DAB Advanced Chromogenic Kit

Catalog No. 501125061
Change view
Click to view available options
Quantity:
50 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125061 50 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Reagent A (DAB Chromogen) is a suspected carcinogen, avoid contact with skin and eyes and dispose in accordance with local regulations. Reagent B (DAB Diluent) contains hydrogen peroxide solution, avoid contact with skin and eyes.
Catalog No. 501125061 Supplier Invitrogen™ Supplier No. 8801496572

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Reagent A (DAB Chromogen) is a suspected carcinogen, avoid contact with skin and eyes and dispose in accordance with local regulations. Reagent B (DAB Diluent) contains hydrogen peroxide solution, avoid contact with skin and eyes.

Includes

  • Reagent A (DAB Chromogen)
  • Reagent B (DAB Diluent)
  • Labeled Empty Dropper Bottle : for storage and application of working DAB solution

Contains the necessary reagents to prepare a DAB substrate working solution.

  • The enzyme peroxidase uses the DAB substrate to produce a brownish-red precipitate
  • These precipiates are insoluble in organic solvent
  • This enzyme/substrate reaction can be used on tissue sections or cells (grown on culture slides or cytospins) to visualize the localization of an HRP-conjugated antibody

Specifications

Detection Method Colorimetric
Quantity 50 tests
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase) Substrate
Product Line eBioscience
Product Type DAB Advanced Chromogenic Kit

For Research Use Only. Not for use in diagnostic procedures.