missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ DA-7645 Detergent

DA-7645, a proprietary blend of surfactants and solvents, enhances manual cleaning in pharmaceutical, biotech, and cosmetic industries. Its mild pH ensures operator safety, while comprehensive validation support makes it easy to use.

$639.43 - $5,310.30

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 8.9 (Diluted)
Scent Unscented
View More Specs

Products 2
Catalog Number Mfr. No. Size Container Type Price Quantity & Availability  
Catalog Number Mfr. No. Size Container Type Price Quantity & Availability  
17018312
View Documents Promotion Details
STERIS™
116505
5 gal. Pail
Each for $639.43
Only null left
 
17018313
View Documents Promotion Details
STERIS™
116501
55 gal. Drum
Each for $5,310.30
Only null left
 
Description

Description

DA-7645 Process and Research Cleaner is specially formulated to enhance manual cleaning with enhanced substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

HOW DA-7645 WORKS

DA-7645 Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms in manual cleaning applications.

WHY DA-7645?

  • Efficacy - Mild pH formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety.
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
  • Productivity - Delivers efficiency to manual cleaning operations using a blend of surfactants, solvents, and other performance-enhancing ingredients.
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Specifications

Specifications

Detergent
Clear Liquid
Unscented
No
8.9 (Diluted)
Non-sterile
Videos
SDS
Documents

Documents