missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STERIS™ DA-7645 Detergent
Description
DA-7645 Process and Research Cleaner is specially formulated to enhance manual cleaning with enhanced substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.
HOW DA-7645 WORKS
DA-7645 Detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms in manual cleaning applications.
WHY DA-7645?
- Efficacy - Mild pH formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety.
- Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
- Productivity - Delivers efficiency to manual cleaning operations using a blend of surfactants, solvents, and other performance-enhancing ingredients.
- Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Specifications
Specifications
| Product Type | Detergent |
| Color-coded | No |
| Form | Clear Liquid |
| pH | 8.9 (Diluted) |
| Scent | Unscented |
| Sterility | Non-sterile |
| Container Type | Drum |
| Size | 55 gal. |