missing translation for 'onlineSavingsMsg'
Learn More

D5 Cyclomethicone Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019451
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019451 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019451 Supplier Merck Emd Millipore Supplier No. PHR1566500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

D5 Cyclomethicone is a cyclic siloxane, that has a silicon-oxygen bond in a cyclic arrangement and methyl groups attached with the silicon atom. It is used in the production of some silicon-based polymers that are widely used in various personal care products.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 541-02-6
Density 0.958 g/mL (at 25°C)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C10H30O5 Si5
MDL Number MFCD00046966
Boiling Point 90°C (10 mmHg)
Synonym Decamethylcyclopentasiloxane
Type Certified Reference Material
Quantity 500 mg
Formula Weight 370.77
Grade Certified Reference Material
Show More Show Less