missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Valinol, 98%

CAS: 4276-09-9 | C5H13NO | 103.17 g/mol

Supplier:  Thermo Scientific Chemicals L1416606

 View more versions of this product

Catalog No. AAL1416606

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

D-(-)-Valinol
1.455
189°C to 190°C
5 g
−11° (c=10 in Water)
C5H13NO
1719138
NWYYWIJOWOLJNR-YFKPBYRVSA-N
(2R)-2-amino-3-methylbutan-1-ol
6950587
98%
4276-09-9
0.931
30°C to 34°C
78°C (172°F)
Air sensitive
MFCD00064297
d-valinol, r---2-amino-3-methyl-1-butanol, r-2-amino-3-methylbutan-1-ol, 2r-2-amino-3-methylbutan-1-ol, r-2-amino-3-methyl-1-butanol, d-2-amino-3-methyl-1-butanol, 2r-2-amino-3-methyl-1-butanol, 1r-1-isopropyl-2-hydroxyethylamine, d---valinol r---2-amino-3-methyl-1-butanol, d-valinol r---2-amino-3-methyl-1-butanol
CC(C)[C@@H](N)CO
103.17
103.17

Chemical Identifiers

4276-09-9
103.17
NWYYWIJOWOLJNR-YFKPBYRVSA-N
6950587
CC(C)[C@@H](N)CO
C5H13NO
MFCD00064297
d-valinol, r---2-amino-3-methyl-1-butanol, r-2-amino-3-methylbutan-1-ol, 2r-2-amino-3-methylbutan-1-ol, r-2-amino-3-methyl-1-butanol, d-2-amino-3-methyl-1-butanol, 2r-2-amino-3-methyl-1-butanol, 1r-1-isopropyl-2-hydroxyethylamine, d---valinol r---2-amino-3-methyl-1-butanol, d-valinol r---2-amino-3-methyl-1-butanol
(2R)-2-amino-3-methylbutan-1-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only