missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Valinol, 98%

CAS: 4276-09-9 | C5H13NO | 103.17 g/mol

$103.59 - $334.26

Chemical Identifiers

CAS 4276-09-9
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.17
MDL Number MFCD00064297
InChI Key NWYYWIJOWOLJNR-YFKPBYRVSA-N
Synonym d-valinol, r---2-amino-3-methyl-1-butanol, r-2-amino-3-methylbutan-1-ol, 2r-2-amino-3-methylbutan-1-ol, r-2-amino-3-methyl-1-butanol, d-2-amino-3-methyl-1-butanol, 2r-2-amino-3-methyl-1-butanol, 1r-1-isopropyl-2-hydroxyethylamine, d---valinol r---2-amino-3-methyl-1-butanol, d-valinol r---2-amino-3-methyl-1-butanol
PubChem CID 6950587
IUPAC Name (2R)-2-amino-3-methylbutan-1-ol
SMILES CC(C)[C@@H](N)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAL1416603
View Documents Promotion Details
N/A 1 g
Each for $103.59
Only null left
 
AAL1416606
View Documents Promotion Details
N/A 5 g
Each for $334.26
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4276-09-9
103.17
NWYYWIJOWOLJNR-YFKPBYRVSA-N
6950587
CC(C)[C@@H](N)CO
C5H13NO
MFCD00064297
d-valinol, r---2-amino-3-methyl-1-butanol, r-2-amino-3-methylbutan-1-ol, 2r-2-amino-3-methylbutan-1-ol, r-2-amino-3-methyl-1-butanol, d-2-amino-3-methyl-1-butanol, 2r-2-amino-3-methyl-1-butanol, 1r-1-isopropyl-2-hydroxyethylamine, d---valinol r---2-amino-3-methyl-1-butanol, d-valinol r---2-amino-3-methyl-1-butanol
(2R)-2-amino-3-methylbutan-1-ol

Specifications

4276-09-9
0.931
30°C to 34°C
−11° (c=10 in Water)
C5H13NO
1719138
NWYYWIJOWOLJNR-YFKPBYRVSA-N
(2R)-2-amino-3-methylbutan-1-ol
6950587
98%
1.455
189°C to 190°C
78°C (172°F)
Air sensitive
MFCD00064297
d-valinol, r---2-amino-3-methyl-1-butanol, r-2-amino-3-methylbutan-1-ol, 2r-2-amino-3-methylbutan-1-ol, r-2-amino-3-methyl-1-butanol, d-2-amino-3-methyl-1-butanol, 2r-2-amino-3-methyl-1-butanol, 1r-1-isopropyl-2-hydroxyethylamine, d---valinol r---2-amino-3-methyl-1-butanol, d-valinol r---2-amino-3-methyl-1-butanol
CC(C)[C@@H](N)CO
103.17
103.17
D-(-)-Valinol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only