missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Valine Methyl Ester Hydrochloride 98.0+%, TCI America™
$69.31 - $188.67
Chemical Identifiers
| CAS | 7146-15-8 |
|---|---|
| Molecular Formula | C6H14ClNO2 |
| Molecular Weight (g/mol) | 167.63 |
| MDL Number | MFCD00237309 |
| InChI Key | KUGLDBMQKZTXPW-NUBCRITNSA-N |
| Synonym | d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl |
| PubChem CID | 11984198 |
| IUPAC Name | methyl (2R)-2-amino-3-methylbutanoate hydrochloride |
| SMILES | Cl.COC(=O)[C@H](N)C(C)C |
Chemical Identifiers
| 7146-15-8 | |
| 167.63 | |
| KUGLDBMQKZTXPW-NUBCRITNSA-N | |
| 11984198 | |
| Cl.COC(=O)[C@H](N)C(C)C |
| C6H14ClNO2 | |
| MFCD00237309 | |
| d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl | |
| methyl (2R)-2-amino-3-methylbutanoate hydrochloride |
Specifications
| 165°C | |
| White | |
| MFCD00237309 | |
| KUGLDBMQKZTXPW-NUBCRITNSA-N | |
| methyl (2R)-2-amino-3-methylbutanoate hydrochloride | |
| 167.63 | |
| 167.63 | |
| Crystalline Powder |
| 7146-15-8 | |
| C6H14ClNO2 | |
| d-valine methyl ester hydrochloride, methyl d-valinate hydrochloride, h-d-val-ome.hcl, r-methyl 2-amino-3-methylbutanoate hydrochloride, d-valinemethyl ester hydrochloride, methyl 2r-2-amino-3-methylbutanoate hydrochloride, d-valine, methyl ester, hydrochloride, h-d-val-ome, methyl d-valinate hcl, h-d-val-ome hcl | |
| Cl.COC(=O)[C@H](N)C(C)C | |
| 1 g | |
| 11984198 | |
| ≥98.0% (N) | |
| D-Valine Methyl Ester Hydrochloride |
Safety and Handling
TSCA : No