accessibility menu, dialog, popup

UserName

H3C(R)OCH3NH2CH3OHCl

CAS RN 7146-15-8

IUPAC Name: methyl (2R)-2-amino-3-methylbutanoate hydrochloride
Synonyms: methyl (2R)-2-amino-3-methylbutanoate hydrochloride
Molecular Weight (g/mol): 167.63
Molecular Formula: C6H14ClNO2
InChi Key: KUGLDBMQKZTXPW-NUBCRITNSA-N
SMILES: Cl.COC(=O)[C@H](N)C(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)